Lauric acid 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dodecanoic acid, Dodecylic acid
16728-0010
16728-0050
16728-0051
16728-1000
16728-5000
TS16728-0010EA
161
USD
TS16728-0010
TS16728-0050
TS16728-0051
TS16728-1000
TS16728-5000
Lauric acid 99%
Lauric acid
Formula:
C₁₂H₂₄O₂ MW: 200.32 g/mol Boiling Pt: 225 °C (100 mmHg) Melting Pt: 44…46 °C Flash Pt: 156 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002736 CAS Number: 143-07-7 EINECS: 205-582-1 Merck Index: 13,05400 |
Specification Test Results
Appearance | White Crystalline powder or flakes |
Infrared spectrum | Conforms |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...