Capric acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Decylic acid, Decanoic acid
16727-0010
16727-0025
16727-1000
16727-5000
TS16727-0010EA
154
USD
TS16727-0010
TS16727-0025
TS16727-1000
TS16727-5000
Capric acid 99%
Capric acid
Formula:
C₁₀H₂₀O₂ MW: 172.27 g/mol Boiling Pt: 268…270 °C (760 mmHg) Melting Pt: 30…32 °C Density: 0.89 g/cm³ Flash Pt: 150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004441 CAS Number: 334-48-5 EINECS: 206-376-4 Merck Index: 13,01764 |
Specification Test Results
Appearance | Colorless or white to cream Crystals and/or chunks or fused solid or liquid |
Infrared spectrum | Conforms |
Melting point | 29 to 34 °C |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...