Ethylenediaminetetraacetic acid disodium magnesium salt hydrate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Ethylenediaminetetraacetic acid disodium magnesium salt xhydrate, EDTA magnesium disodium salt hydrate, EDTA-Na2Mg salt
45793-0250
45793-1000
45793-5000
TS45793-0250EA
24
USD
TS45793-0250
TS45793-1000
TS45793-5000
Ethylenediaminetetraacetic acid disodium magnesium salt hydrate 97%
Ethylenediaminetetraacetic acid disodium magnesium salt hydrate
Formula:
C₁₀H₁₂MgN₂Na₂O₈·xH₂O MW: 358.50 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00078217 CAS Number: 194491-32-2 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Assay | ≥96.0 % (on dry substance) |
Loss on drying | 10 % to 20 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...