Azomethine H monosodium salt monohydrate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxy-5-(2-hydroxybenzylideneamino)naphthalene-2,7-disulphonic acid monosodium salt

45709-0010 45709-0050 45709-0250
TS45709-0010EA 20 USD
TS45709-0010 TS45709-0050 TS45709-0250
Azomethine H monosodium salt monohydrate 95%
Azomethine H monosodium salt monohydrate
Formula: C₁₇H₁₂NNaO₈S₂·H₂O
MW: 463.41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149588
CAS Number: 304655-82-1

Order Now

Specification Test Results

Appearance Yellow to orange Powder
Infrared spectrum Conforms
Water ≤6 %
Titration with NaOH ≥94 %
Sodium (Na) 4.5 to 5.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR