1,4-Diazabicyclo[2.2.2]octanebis(sulfur dioxide)adduct 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: DABSO, 1,4-Diazabicyclo[2.2.2]octane-1,4-diium-1,4-disulphinate, DABCO-Bis(sulphur dioxide), Bis(sulphur dioxide)-1,4-diazabicyclo[2.2.2]octane

45635-0010 45635-0050
TS45635-0010EA 52 USD
TS45635-0010 TS45635-0050
1,4-Diazabicyclo[2.2.2]octanebis(sulfur dioxide)adduct 97%
1,4-Diazabicyclo[2.2.2]octanebis(sulfur dioxide)adduct
Formula: C₆H₁₂N₂O₄S₂
MW: 240.29 g/mol
Melting Pt: 168…172 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22377802
CAS Number: 119752-83-9
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 168 to 172 °C
Assay ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR