p-Toluenesulfonyl cyanide 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Methylbenzenesulphonyl cyanide, Tosyl cyanide

45582-0010 45582-0050
TS45582-0010EA 69 USD
TS45582-0010 TS45582-0050
p-Toluenesulfonyl cyanide 95%
p-Toluenesulfonyl cyanide
Formula: C₈H₇NO₂S
MW: 181.21 g/mol
Melting Pt: 47…50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009977
CAS Number: 19158-51-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to tan Crystalline powder
Infrared spectrum Conforms
Melting point 47 to 50 °C
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR