3-Diethylamino-1-propyne 97%

Supplier: Thermo Scientific Chemicals
Danger

45558-0050 45558-0250 45558-1000
TS45558-0050EA 37 USD
TS45558-0050 TS45558-0250 TS45558-1000
3-Diethylamino-1-propyne 97%
3-Diethylamino-1-propyne
Formula: C₇H₁₃N
Boiling Pt: 118…120 °C
Density: 0.811 g/cm³
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00039881
CAS Number: 4079-68-9
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR