Ethyl viologen dibromide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,1'-Diethyl-[4,4'-bipyridine]-1,1'-diium bromide, 1,1'-Diethyl-4,4'-bipyridinium dibromide

45503-0010 45503-0050
TS45503-0010EA 41 USD
TS45503-0010 TS45503-0050
Ethyl viologen dibromide 97%
Ethyl viologen dibromide
Formula: C₁₄H₁₈Br₂N₂
Melting Pt: 278 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00134590
CAS Number: 53721-12-3

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR