Sodium trifluoromethanesulfinate 98%

Supplier: Thermo Scientific Chemicals
Warning

45499-0010 45499-0050 45499-0250
TS45499-0010EA 38 USD
TS45499-0010 TS45499-0050 TS45499-0250
Sodium trifluoromethanesulfinate 98%
Sodium trifluoromethanesulfinate
Formula: CHF₃O₂S
Melting Pt: 350 °C
Storage Temperature: Ambient
MDL Number: MFCD03092989
CAS Number: 2926-29-6

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤1 %
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR