2,2,2-Trifluoroethyl trifluoromethanesulfonate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,2,2-Trifluoroethyltrifluoromethanesulphonate

45497-0010 45497-0050 45497-0250
TS45497-0010EA 60 USD
TS45497-0010 TS45497-0050 TS45497-0250
2,2,2-Trifluoroethyl trifluoromethanesulfonate 97%
2,2,2-Trifluoroethyl trifluoromethanesulphonate
Formula: C₃H₂F₆O₃S
MW: 232.10 g/mol
Boiling Pt: 89…91 °C (740 mmHg)
Density: 1.61 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00671579
CAS Number: 6226-25-1
UN: 2927
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR