4,4'-Dibromo-2,2'-bipyridyl 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,4'-Dibromo-2,2'-bipyridine

45490-0010 45490-0050
TS45490-0010EA 120 USD
TS45490-0010 TS45490-0050
4,4'-Dibromo-2,2'-bipyridyl 98%
4,4'-Dibromo-2,2'-bipyridyl
Formula: C₁₀H₆Br₂N₂
MW: 313.98 g/mol
Melting Pt: 140…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00667763
CAS Number: 18511-71-2

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 140 to 145 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR