(Isocyanoimino)triphenylphosphorane 90%

Supplier: Thermo Scientific Chemicals
Warning

45292-0010 45292-0050
TS45292-0010EA 73 USD
TS45292-0010 TS45292-0050
(Isocyanoimino)triphenylphosphorane 90%
(Isocyanoimino)triphenylphosphorane
Formula: C₁₉H₁₅N₂P
Melting Pt: 160…161 °C
Storage Temperature: Ambient
MDL Number: MFCD09038528
CAS Number: 73789-56-7

Order Now

Specification Test Results

Appearance White to light yellow-orange Crystalline powder
Infrared spectrum Conforms
HPLC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR