Xylenol orange tetrasodium salt ACS

Supplier: Thermo Scientific Chemicals

44760-0010 44760-0050 44760-0250
TS44760-0010EA 36 USD
TS44760-0010 TS44760-0050 TS44760-0250
Xylenol orange tetrasodium salt ACS
Xylenol orange tetrasodium salt
Formula: C₃₁H₂₈N₂Na₄O₁₃S
Melting Pt: 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00044293
CAS Number: 3618-43-7

Order Now

Specification Test Results

Appearance Red-brown to dark brown Crystalline powder
Suitability (for Zn titration) Passes test
Clarity of solution Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR