Crystal violet ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
44757-0500
44757-2500
TS44757-0500EA
67
USD
TS44757-0500
TS44757-2500
Crystal violet ACS
Crystal violet
Formula:
C₂₅H₃₀ClN₃ Melting Pt: 173 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011750 CAS Number: 548-62-9 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | Dark green Powder or crystals |
Assay | ≥90.0 % (on dry substance) |
Loss on drying | ≤7.5 % |
Absorbance characteristics | (0.98-1.9) Passes test |
Sensitivity | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...