2',7'-Dichlorofluorescein ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2',7'-Dichloro-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

44693-0010 44693-0100
TS44693-0010EA 42 USD
TS44693-0010 TS44693-0100
2',7'-Dichlorofluorescein ACS
2',7'-Dichlorofluorescein
Formula: C₂₀H₁₀Cl₂O₅
MW: 401.20 g/mol
Melting Pt: 280 °C (dec.)
Density: 0.79 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00005047
CAS Number: 76-54-0

Order Now

Specification Test Results

Appearance Light orange to red-brown Powder
Suitability (as adsorption indicator) Passes test
Clarity of alcohol solution Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR