Bis(2-bromoethyl)ether 90%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Bromo-2-(2-bromoethoxy)ethane, 2-Bromoethyl ether

44586-0050 44586-0250
TS44586-0050EA 80 USD
TS44586-0050 TS44586-0250
Bis(2-bromoethyl)ether 90%
Bis(2-bromoethyl)ether
Formula: C₄H₈Br₂O
MW: 231.93 g/mol
Boiling Pt: 92…93 °C (12 mmHg)
Density: 1.845 g/cm³
Flash Pt: 85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039196
CAS Number: 5414-19-7

Order Now

Specification Test Results

Appearance Colorless to brown Liquid (May darken on storage)
Infrared spectrum Conforms
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR