(7-Azabenzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate, (7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate, (3-Hydroxy-3H-1,2,3-triazolo[4,5-b]pyridinato-O)tri-1-pyrrolidinylphosphonium hexafluorophosphate, ((3H-[1,2,3]Triazolo[4,5-b]pyridin-3-yl)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V), PyAOP, (3-Hydroxy-3H-1,2,3-triazolo[4,5-b]pyridinato-O)tri-1-pyrrolidinylphosphorous hexafluorophosphate
44061-0010
44061-0050
TS44061-0010EA
94
USD
TS44061-0010
TS44061-0050
(7-Azabenzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) 98%
(7-Azabenzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)
Formula:
C₁₇H₂₇F₆N₇OP₂ MW: 521.4 g/mol Melting Pt: 153…155 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03703417 CAS Number: 156311-83-0 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Elemental Analysis | 18.33 to 19.28 % (N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...