N,N,N',N'-Bis(tetramethylene)-O-(N-succinimidyl)uronium hexafluorophosphate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(((2,5-Dioxopyrrolidin-1-yl)oxy)(pyrrolidin-1-yl)methylene)pyrrolidin-1-ium hexafluorophosphate(V), HSPyU, Dipyrrolidino(N-succinimidyloxy)carbenium Hexafluorophosphate, Dipyrrolidino(N-succinimidyloxy)carbenium hexafluorophosphate

44059-0050 44059-0250
TS44059-0050EA 214 USD
TS44059-0050 TS44059-0250
N,N,N',N'-Bis(tetramethylene)-O-(N-succinimidyl)uronium hexafluorophosphate 97%
N,N,N',N'-Bis(tetramethylene)-O-(N-succinimidyl)uronium hexafluorophosphate
Formula: C₁₃H₂₀F₆N₃O₃P
MW: 411.29 g/mol
Melting Pt: 141…143.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00192457
CAS Number: 207683-26-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Elemental Analysis 9.81 to 10.63 % (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR