2-Chloro-1,3-dimethylimidazolinium chloride 90%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
DMC
43952-0050
43952-0250
43952-1000
TS43952-0050EA
63
USD
TS43952-0050
TS43952-0250
TS43952-1000
2-Chloro-1,3-dimethylimidazolinium chloride 90%
2-Chloro-1,3-dimethylimidazolinium chloride
Formula:
C₅H₁₀Cl₂N₂ MW: 169.05 g/mol Melting Pt: 112…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD09039290 CAS Number: 37091-73-9 |
Specification Test Results
Appearance | White to off-white to brown Crystalline powder or solid |
Infrared spectrum | Conforms |
HPLC | ≥88.0 % |
Elemental Analysis | 14.58 to 18.56 % (N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...