COMU 97%

Supplier: Thermo Scientific Chemicals
Warning

43948-0050 43948-0100 43948-1000
TS43948-0050EA 64 USD
TS43948-0050 TS43948-0100 TS43948-1000
COMU 97%
COMU
Formula: C₁₂H₁₉F₆N₄O₄P
Melting Pt: 151.4…152.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11975052
CAS Number: 1075198-30-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point ≥145.0 °C (Dec)
Water ≤3.00 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR