Eriochrome Black T ACS indicator grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Naphthalenesulphonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1), Eriochrome black T, Indicator, EBT, Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulphonate, Solochrome Black T
43914-0010
43914-2500
TS43914-0010EA
177
USD
TS43914-0010
TS43914-2500
Eriochrome Black T ACS indicator grade
Eriochrome Black T
Formula:
C₂₀H₁₂N₃NaO₇S MW: 461.39 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003935 CAS Number: 1787-61-7 |
Specification Test Results
Appearance | Brown to brown-black Powder |
Infrared spectrum | Conforms |
Suitability | (as complexom. indicator) Passes test |
Clarity of solution | (0.1% water) Pases test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...