1,4-Diazabicyclo[3.2.2]nonane 95%

Supplier: Thermo Scientific Chemicals
Danger

43890-0010 43890-0050
TS43890-0010EA 274 USD
TS43890-0010 TS43890-0050
1,4-Diazabicyclo[3.2.2]nonane 95%
1,4-Diazabicyclo[3.2.2]nonane
Formula: C₇H₁₄N₂
Boiling Pt: 77…79 °C (6Torr)
Melting Pt: 58…79 °C
Storage Temperature: Ambient
MDL Number: MFCD14581694
CAS Number: 283-38-5
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to brown Liquid or waxy solid
Infrared spectrum Conforms
GC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR