Tetrakis(decyl)ammonium bromide 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Tetrakis(decyl)ammonium bromide, IPC-TDA-Br, Tetra(decyl)ammonium bromide
43567-0100
43567-0500
TS43567-0100EA
141
USD
TS43567-0100
TS43567-0500
Tetrakis(decyl)ammonium bromide 98%
Tetrakis(decyl)ammonium bromide
Formula:
C₄₀H₈₄BrN MW: 659.0057 g/mol Melting Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043166 CAS Number: 14937-42-9 |
Specification Test Results
Appearance | White to light beige Powder |
Infrared spectrum | Conforms |
Melting point | 86 to 90 °C |
Titration Argentometric | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...