4,7-Dimethoxy-1,10-phenanthroline 97%

Supplier: Thermo Scientific Chemicals
Danger

43378-0010 43378-2500
TS43378-0010EA 343 USD
TS43378-0010 TS43378-2500
4,7-Dimethoxy-1,10-phenanthroline 97%
4,7-Dimethoxy-1,10-phenanthroline
Formula: C₁₄H₁₂N₂O₂
MW: 240.2573 g/mol
Melting Pt: 212…214 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00233883
CAS Number: 92149-07-0
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to brown Crystalline powder or solid
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR