Vanadium(III) chloride tetrahydrofuran complex (1:3) 0.5M in methylene chloride, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trichlorotris(tetrahydrofuran)vanadium, Vanadium(III) chloride tetrahydrofuran adduct, Trichlorotris(tetrahydrofuran)vanadium(III)

43366-1000 43366-8000
TS43366-1000EA 139 USD
TS43366-1000 TS43366-8000
Vanadium(III) chloride tetrahydrofuran complex (1:3) 0.5M in methylene chloride, AcroSeal™
Vanadium(III) chloride tetrahydrofuran complex (1:3)
Formula: VCl₃·3C₄H₈O
MW: 373.62 g/mol
Density: 1.312 g/cm³
Flash Pt: –8 °C
Storage Temperature: Ambient
MDL Number: MFCD00145515
CAS Number: 19559-06-9
UN: 1593
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light red to dark red Solution
Titration Argentometric 0.40 to 0.60 M
Water ≤0.005 %
GC ≥99.4 %
Stabilizer Approx. 50 ppm BHT
Peroxide value ≤50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR