Trimethyl orthoformate, anhydrous 99+%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

42928-1000 42928-8000
TS42928-1000EA 40 USD
TS42928-1000 TS42928-8000
Trimethyl orthoformate, anhydrous 99+%, AcroSeal™
Trimethyl orthoformate
Formula: C₄H₁₀O₃
Boiling Pt: 101…102 °C (760 mmHg)
Melting Pt: –53 °C
Density: 0.97 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00008483
CAS Number: 149-73-5
UN: 3271
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.0 %
Residue after evaporation ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR