Triethyl orthoformate, anhydrous 98%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Warning

42927-1000 42927-8000
TS42927-1000EA 49 USD
TS42927-1000 TS42927-8000
Triethyl orthoformate, anhydrous 98%, AcroSeal™
Triethyl orthoformate
Formula: C₇H₁₆O₃
MW: 148.2 g/mol
Boiling Pt: 146 °C (760 mmHg)
Melting Pt: –61 °C
Density: 0.89 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00009230
CAS Number: 122-51-0
UN: 2524
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥97.5 %
Residue after evaporation ≤0.005 %
Acidity ≤0.0001 meq/g
Substance darkened by H2SO4 ≤10 APHA
Calcium (Ca) ≤0.5 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.02 ppm
Magnesium (Mg) ≤0.05 ppm
Potassium (K) ≤0.1 ppm
Sodium (Na) ≤0.5 ppm
Zinc (Zn) ≤0.2 ppm
Total phosphorus (P) ≤0.1 ppm
Total silicon (Si) ≤0.05 ppm
Total sulfur (S) ≤0.001 %
Bromine number ≤0.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR