Diazolidinyl urea 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3-Bis(hydroxymethyl)-5-[1,3-bis(hydroxymethyl)ureido]hydantoin, 1-(1,3-Bis(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl)-1,3-bis(hydroxymethyl)urea

42858-0250 42858-2500
TS42858-0250EA 43 USD
TS42858-0250 TS42858-2500
Diazolidinyl urea 95%
Diazolidinyl urea
Formula: C₈H₁₄N₄O₇
MW: 278.22 g/mol
Melting Pt: 227…228 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03547942
CAS Number: 78491-02-8

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤3 %
Solubility (50 mg/ml in water) Clear colorless
Elemental Analysis 19.0 to 21.5 % N
Hydroxyl value 102 to 124 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR