EGTA-Na4 (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt) 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,12-Bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecane-1,14-dioic acid, tetrasodium salt, Ethylene glycol-bis(beta-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt, EGTA tetrasodium

42857-0100 42857-0500
TS42857-0100EA 98 USD
TS42857-0100 TS42857-0500
EGTA-Na4 (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt) 95%
EGTA-Na4 (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt)
Formula: C₁₄H₂₀N₂Na₄O₁₀
MW: 468.28 g/mol
Melting Pt: 54…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00274420
CAS Number: 13368-13-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Assay 18.46 to 20.82 % (ICP on dried sample)
Loss on drying ≤20 % (110°C, 16 hrs)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR