N-Methyldicyclohexylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

42839-0010 42839-2500
TS42839-0010EA 198 USD
TS42839-0010 TS42839-2500
N-Methyldicyclohexylamine 97%
N-Methyldicyclohexylamine
Formula: C₁₃H₂₅N
Boiling Pt: 265 °C (760 mmHg)
Density: 0.921 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00014289
CAS Number: 7560-83-0
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4890 to 1.4910 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR