(S)-(-)-Menthyltoluene-4-sulfinate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (1R,2S,5R)-(-)-Menthyl (S)-p-toluenesulphinate, (S)-((1R,2S,5R)-2-isopropyl-5-methylcyclohexyl) 4-methylbenzenesulphinate

42811-0010 42811-0100
TS42811-0010EA 36 USD
TS42811-0010 TS42811-0100
(S)-(-)-Menthyltoluene-4-sulfinate 98%
(S)-(-)-Menthyltoluene-4-sulphinate
Formula: C₁₇H₂₆O₂S
MW: 294.4521 g/mol
Melting Pt: 102…104 °C
Storage Temperature: Freezer
MDL Number: MFCD00010192
CAS Number: 1517-82-4

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 100 to 107 °C
HPLC ≥97.5 %
Specific optical rotation -190° to -204° (20 °C, 589 nm) (c=2,acetone)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR