3-(Benzyldimethylammonio)propanesulfonate 95%

Supplier: Thermo Scientific Chemicals

Synonyms: NDSB-256, 3-(Benzyldimethylammonio) propane-1-sulphonate, 3-[dimethylbenzylamino]propanesulphonic acid

42807-0050 42807-0250
TS42807-0050EA 66 USD
TS42807-0050 TS42807-0250
3-(Benzyldimethylammonio)propanesulfonate 95%
3-(Benzyldimethylammonio)propanesulphonate
Formula: C₁₂H₁₉NO₃S
MW: 257.348 g/mol
Melting Pt: 245…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00225018
CAS Number: 81239-45-4

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
NMR ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR