N-Succinimidyl Bromoacetate 90%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,5-Dioxopyrrolidin-1-yl 2-bromoacetate, N-(Bromoacetoxy)succinimide, Bromoacetic Acid N-Succinimidyl Ester, Bromoacetic acid N-hydroxysuccinimide ester

42778-0010 42778-0050 42778-2500
TS42778-0010EA 857 USD
TS42778-0010 TS42778-0050 TS42778-2500
N-Succinimidyl Bromoacetate 90%
N-Succinimidyl Bromoacetate
Formula: C₆H₆BrNO₄
MW: 236.02 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00058571
CAS Number: 42014-51-7

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Elemental Analysis ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR