5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-(Trifluoromethyl)-5H-dibenzo[b,d]thiophen-5-ium trifluoromethanesulphonate

42767-0010 42767-0050
TS42767-0010EA 117 USD
TS42767-0010 TS42767-0050
5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate 96%
5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate
Formula: C₁₄H₈F₆O₃S₂
MW: 402.325 g/mol
Melting Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00236132
CAS Number: 129946-88-9

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder or solid
Infrared spectrum Conforms
HPLC ≥95 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR