4-(Diphenylphosphino)benzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

42653-0010 42653-0050
TS42653-0010EA 82 USD
TS42653-0010 TS42653-0050
4-(Diphenylphosphino)benzoic acid 97%
4-(Diphenylphosphino)benzoic acid
Formula: C₁₉H₁₅O₂P
Melting Pt: 157…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00407264
CAS Number: 2129-31-9

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 155 to 160 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR