Thiourea 99+% ACS

Supplier: Thermo Scientific Chemicals
Warning

42454-0010 42454-2500
TS42454-0010EA 301.47 USD
TS42454-0010 TS42454-2500
Thiourea 99+% ACS
Thiourea
Formula: NH₂CSNH₂
Melting Pt: 171…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00008067
CAS Number: 62-56-6
EINECS: 200-543-5
UN: 3077
ADR: 9,III
Merck Index: 13,09443
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fff6222c54fe16ea4d36edcbf320745cb1e18d6&vid=e9ae5dc49e92d68f893244a5cd4e0245f289ed09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 174 to 177 °C
Titration Argentometric ≥99.0 % (On dried basis)
Loss on drying ≤0.5 % (1.5 g, 105 °C, 2 h)
Solubility (16.5g in 100ml water,25 °C) Passes test
Residue after ignition ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR