Methyl red ACS
Supplier: Thermo Scientific Chemicals
Synonyms:
2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
42397-0100
42397-0250
42397-1000
TS42397-0100EA
32
USD
TS42397-0100
TS42397-0250
TS42397-1000
Methyl red ACS
Methyl red
Formula:
C₁₅H₁₅N₃O₂ MW: 269.3 g/mol Melting Pt: 178…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002425 CAS Number: 493-52-7 EINECS: 207-776-1 Merck Index: 12,06199 |
Specification Test Results
Appearance | Dark red to brown to purple Powder |
Infrared spectrum | Conforms |
Melting point | 179 to 182 °C |
Visual transition interval | From pH 4.2 (pink) to pH 6.2 (yellow) |
Clarity of alcohol solution | Passes test |
Specific optical rotation | +52.0° to +62.0° (20 °C, 589 nm) (neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...