Vanadium(IV) oxide phthalocyanine 85%, Practical grade

Supplier: Thermo Scientific Chemicals
Warning

42249-0250 42249-1000
TS42249-0250EA 326 USD
TS42249-0250 TS42249-1000
Vanadium(IV) oxide phthalocyanine 85%, Practical grade
Vanadium(IV) oxide phthalocyanine
Formula: C₃₂H₁₆N₈OV
Storage Temperature: Ambient
MDL Number: MFCD00041966
CAS Number: 13930-88-6
UN: 3285
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Dark blue to purple Powder
Infrared spectrum Conforms
Assay ≥80 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR