Tris(4-methylphenyl)phosphine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tri-p-tolylphosphine, Tris(p-tolyl)phosphine

42233-0010 42233-0050
TS42233-0010EA 26 USD
TS42233-0010 TS42233-0050
Tris(4-methylphenyl)phosphine 98%
Tris(4-methylphenyl)phosphine
Formula: C₂₁H₂₁P
MW: 304.37 g/mol
Melting Pt: 145…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00008542
CAS Number: 1038-95-5
EINECS: 213-863-5

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 144 to 148 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR