Tris(4-methoxyphenyl)phosphine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tris(p-methoxyphenyl)phosphine

42224-0010 42224-0100
TS42224-0010EA 43 USD
TS42224-0010 TS42224-0100
Tris(4-methoxyphenyl)phosphine 95%
Tris(4-methoxyphenyl)phosphine
Formula: C₂₁H₂₁O₃P
MW: 352.37 g/mol
Melting Pt: 131…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00014896
CAS Number: 855-38-9
EINECS: 212-723-0

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 131 to 134 °C
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR