Symclosene 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trichloro-1,3,5-triazinetrion, 1,3,5-Trichlorohexahydro-1,3,5-triazine-2,4,6-trione, Trichlorocyanuric acid, TCCA, 1,3,5-Trichloro-1,3,5-triazine-2,4,6-trione, Trichloroisocyanuric acid

42154-0010 42154-0050 42154-2500
TS42154-0010EA 96 USD
TS42154-0010 TS42154-0050 TS42154-2500
Symclosene 99%
Symclosene
Formula: C₃Cl₃N₃O₃
Melting Pt: 245…251 °C
Storage Temperature: Ambient
MDL Number: MFCD00006553
CAS Number: 87-90-1
EINECS: 201-782-8
UN: 2468
ADR: 5.1,II
Merck Index: 12,09188

Order Now

Specification Test Results

Appearance White to off-white Powder or granules
Infrared spectrum Conforms
Titration Iodometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR