o-Tolidine 95% pract.

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4,4'-Bi-o-toluidine

42112-0250 42112-1000 42112-2500 42112-5000
TS42112-0250EA 61 USD
TS42112-0250 TS42112-1000 TS42112-2500 TS42112-5000
o-Tolidine 95% pract.
o-Tolidine
Formula: [-C₆H₃(CH₃)-₄-NH₂]₂
MW: 212.29 g/mol
Boiling Pt: ∼361 °C (1013 hPa)
Melting Pt: 125…132 °C
Flash Pt: 244 °C
Storage Temperature: Ambient
MDL Number: MFCD00014773
CAS Number: 119-93-7
EINECS: 204-358-0
UN: 3077
ADR: 9,III
Merck Index: 12,09651

Order Now

Specification Test Results

Appearance Beige to grey to brown Powder
Infrared spectrum Conforms
Melting point 125 to 132 °C
Titration with HClO4 ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR