Thymol blue sodium salt ACS

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bis[5-methyl-2-(1-methylethyl)phenolate] S,S-dioxide, Sodium 4-(3-(4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-2-isopropyl-5-methylphenolate, Thymol blue water soluble, Thymol Blue sodium

42094-0100 42094-0250 42094-1000
TS42094-0100EA 62 USD
TS42094-0100 TS42094-0250 TS42094-1000
Thymol blue sodium salt ACS
Thymol blue sodium salt
Formula: C₂₇H₂₉NaO₅S
MW: 488.58 g/mol
Melting Pt: 283 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00151093
CAS Number: 62625-21-2
EINECS: 263-650-6

Order Now

Specification Test Results

Appearance Dark orange to green-black to brown-black Powder
Infrared spectrum Conforms
Visual transition interval From pH 8.0 (yellow) to pH 9.2 (blue)
Clarity of solution Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR