Thymol blue ACS

Supplier: Thermo Scientific Chemicals

Synonyms: Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

42093-0050 42093-0100 42093-1000
TS42093-0050EA 42 USD
TS42093-0050 TS42093-0100 TS42093-1000
Thymol blue ACS
Thymol blue
Formula: C₂₇H₃₀O₅S
Melting Pt: 221 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005869
CAS Number: 76-61-9
EINECS: 200-973-3
Merck Index: 12,09541

Order Now

Specification Test Results

Appearance Dark green to dark brown Crystalline powder
Infrared spectrum Conforms
Visual transition interval from pH 8.0 (yellow) to pH 9.2 (blue)
Clarity of solution passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR