N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N1,N1,N4,N4-Tetramethylbenzene-1,4-diamine dihydrochloride, TMPD 2 HCl, N,N,N',N'-Tetramethyl-1,4-phenylenediamine dihydrochloride, N,N,N',N'-Tetramethyl-1,4-phenylenediammonium dichloride
42060-0050
42060-0250
42060-1000
TS42060-0050EA
62
USD
TS42060-0050
TS42060-0250
TS42060-1000
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride 98+%
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride
Formula:
C₁₀H₁₈Cl₂N₂ MW: 237.17 g/mol Melting Pt: 220…224 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012482 CAS Number: 637-01-4 EINECS: 211-274-8 Merck Index: 12,09367 |
Specification Test Results
Appearance | White to beige to grey to light brown Crystalline powder |
Infrared spectrum | Conforms |
Titration with HClO4 | 98.0 to 102.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...