Safranine O 85%, pure, certified

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: CI 50240

41921-0250 41921-1000
TS41921-0250EA 67 USD
TS41921-0250 TS41921-1000
Safranine O 85%, pure, certified
Safranine O
Formula: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011759
CAS Number: 477-73-6
EINECS: 207-518-8

Order Now

Specification Test Results

Appearance Dark green to dark brown Powder
Infrared spectrum Conforms
Dye content ≥80 %
UV Lambda max 530 to 534 nm
Absorption Ratio P-15/P+15 1.1 to 1.32
BSC Certification Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR