Basic fuchsin 88%, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline
41914-0025
41914-1000
41914-5000
TS41914-0025EA
966
USD
TS41914-0025
TS41914-1000
TS41914-5000
Basic fuchsin 88%, pure
Basic fuchsin
Formula:
C₂₀H₂₀ClN₃ MW: 337.85 g/mol Melting Pt: 250 °C (dec.) Flash Pt: 200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012569 CAS Number: 632-99-5 EINECS: 211-189-6 Merck Index: 12,05686 |
Specification Test Results
Appearance | Green Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤10 % (105°C) |
Dye content | ≥83 % (on dry substance) |
UV Lambda max | 549 to 555 nm (in 50:50 IMS:water) |
Absorption Ratio P-15/P+15 | 1.1 to 1.4 |
Specific absorbance A (1%/1cm) | ≥1800 (at lambda max.) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...