Rhodamine 6G perchlorate 99%, pure laser grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
41903-0025
41903-5000
TS41903-0025EA
185
USD
TS41903-0025
TS41903-5000
Rhodamine 6G perchlorate 99%, pure laser grade
Rhodamine 6G perchlorate
Formula:
C₂₈H₃₁ClN₂O₇ MW: 543.02 g/mol Melting Pt: 263…265 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013117 CAS Number: 13161-28-9 EINECS: 236-099-4 |
Specification Test Results
Appearance | Pink to red Crystalline powder or crystals |
Infrared spectrum | Conforms |
UV spectrum | Complies |
TLC | ≥98.5 % |
UV Lambda max | 528 nm (in Methanol) |
Fluorescence max | 547 nm (in Methanol) |
Optimum lasing | 563 nm (in Methanol) |
Suitability | (for use as a laser dye) Passes test |
Molar absorption coefficient | 104000 l/(mole.cm) (Lambda max) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...