Rhodamine 6G 99%, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
9-(2-(Ethoxycarbonyl)phenyl)-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride, 9-[2-(Ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride
41901-0050
41901-0250
41901-1000
TS41901-0050EA
34
USD
TS41901-0050
TS41901-0250
TS41901-1000
Rhodamine 6G 99%, pure
Rhodamine 6G
Formula:
C₂₈H₃₁ClN₂O₃ MW: 479.02 g/mol Boiling Pt: ∼200 °C (1013 hPa) Melting Pt: 229.4 °C |
MDL Number:
MFCD00012665 CAS Number: 989-38-8 EINECS: 213-584-9 UN: 2811 |
Specification Test Results
Appearance | Red-brown to brown Crystalline powder |
Infrared spectrum | Conforms |
UV spectrophotometry | ≥98.5 % |
Enantiomeric excess | ≥97.5 % |
Specific optical rotation | -33° to -39° (20°C, 589 nm) (c=1, CHCl3) |
Titration with NaOH | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...