Pyronin G, certified

Supplier: Thermo Scientific Chemicals
Warning

41863-0010 41863-0050
TS41863-0010EA 53 USD
TS41863-0010 TS41863-0050
Pyronin G, certified
Pyronin G
Formula: C₁₇H₁₉ClN₂O
Melting Pt: 250…260 °C
Storage Temperature: Ambient
MDL Number: MFCD00011725
CAS Number: 92-32-0
EINECS: 202-147-8
Merck Index: 12,08190

Order Now

Specification Test Results

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
Dye content ≥45 %
UV Lambda max 546 to 551 nm
Absorption Ratio P-15/P+15 1.23 to 1.7
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR